Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-chloro-2,6-dimethoxybenzamide, 97%

Click to view available options
Quantity:
1g
5g
175135-60-1
C9H9BrClNO3
294.529
MFCD00052965
GBVBCQIVBHAEAZ-UHFFFAOYSA-N
maybridge1_001686,acmc-1c868,benzamide,3-bromo-5-chloro-2,6-dimethoxy,3-bromanyl-5-chloranyl-2,6-dimethoxy-benzamide
2800507
3-bromo-5-chloro-2,6-dimethoxybenzamide
COC1=C(C(=C(C=C1Cl)Br)OC)C(=O)N
175135-60-1
C9H9BrClNO3
294.529
MFCD00052965
GBVBCQIVBHAEAZ-UHFFFAOYSA-N
maybridge1_001686,acmc-1c868,benzamide,3-bromo-5-chloro-2,6-dimethoxy,3-bromanyl-5-chloranyl-2,6-dimethoxy-benzamide
2800507
3-bromo-5-chloro-2,6-dimethoxybenzamide
COC1=C(C(=C(C=C1Cl)Br)OC)C(=O)N
Quantity 1g
Formula Weight 294.53
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-chloro-2,6-dimethoxybenzamide