Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-(chloromethyl)pyridine hydrochloride, 96%

Click to view available options
Quantity:
250mg
1g
5g
120277-69-2
C6H5BrClN
206.467
MFCD10697600
NLPHAZLCNNDGPS-UHFFFAOYSA-N
3-bromo-5-chloromethyl pyridine,pyridine, 3-bromo-5-chloromethyl,3-bromo-5-chloromethyl pyridine,5-bromonicotinyl chloride,3-bromo-5-chloromethyl-pyridine,3-bromo-5-chloromethyl-pyridine,pyridine, 3-bromo-5-chloromethyl-9ci
11701259
3-bromo-5-(chloromethyl)pyridine
C1=C(C=NC=C1Br)CCl
120277-69-2
C6H5BrClN
206.467
MFCD10697600
NLPHAZLCNNDGPS-UHFFFAOYSA-N
3-bromo-5-chloromethyl pyridine,pyridine, 3-bromo-5-chloromethyl,3-bromo-5-chloromethyl pyridine,5-bromonicotinyl chloride,3-bromo-5-chloromethyl-pyridine,3-bromo-5-chloromethyl-pyridine,pyridine, 3-bromo-5-chloromethyl-9ci
11701259
3-bromo-5-(chloromethyl)pyridine
C1=C(C=NC=C1Br)CCl
Quantity 250mg
Color White
UN Number UN3261
Beilstein 1562735
Formula Weight 242.93
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Bromo-5-(chloromethyl)pyridine hydrochloride

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.