Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-chloropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
73583-39-8
C5H3BrClN
192.44
BELDOPUBSLPBCQ-UHFFFAOYSA-N
3-chloro-5-bromopyridine,3-bromo-5-chloro-pyridine,pyridine, 3-bromo-5-chloro,5-bromo-3-chloropyridine,pubchem7700,3-brom-5-chlorpyridin,acmc-209orf,#,ksc377a4j,3-bromo-5-chloropyridine
606256
3-bromo-5-chloropyridine
C1=C(C=NC=C1Br)Cl
73583-39-8
C5H3BrClN
192.44
BELDOPUBSLPBCQ-UHFFFAOYSA-N
3-chloro-5-bromopyridine,3-bromo-5-chloro-pyridine,pyridine, 3-bromo-5-chloro,5-bromo-3-chloropyridine,pubchem7700,3-brom-5-chlorpyridin,acmc-209orf,#,ksc377a4j,3-bromo-5-chloropyridine
606256
3-bromo-5-chloropyridine
C1=C(C=NC=C1Br)Cl
Quantity 5g
Packaging Glass bottle
Melting Point 78.7°C to 80.1°C
Color White to Brown
Infrared Spectrum Authentic
Water 3% Max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 192.44
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-chloropyridine
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger