Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-ethoxy-4-hydroxybenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
3111-37-3
C9H9BrO3
245.072
MFCD00173904
GWEFTCNMUHHQLP-UHFFFAOYSA-N
3-bromo-5-ethoxy-4-hydroxy-benzaldehyde,acmc-1cof4,3-bromo-4-hydroxy-5-ethoxybenzaldehyde,3-bromo-5-ethoxy4-hydroxy-benzaldehyde,5-bromo-3-ethoxy-4-hydroxybenzaldehyde,3-bromo-5-ethoxy-4-hydroxy benzaldehyde,benzaldehyde,3-bromo-5-ethoxy-4-hydroxy,3-bromanyl-5-ethoxy-4-oxidanyl-benzaldehyde
612496
3-bromo-5-ethoxy-4-hydroxybenzaldehyde
CCOC1=C(C(=CC(=C1)C=O)Br)O
3111-37-3
C9H9BrO3
245.072
MFCD00173904
GWEFTCNMUHHQLP-UHFFFAOYSA-N
3-bromo-5-ethoxy-4-hydroxy-benzaldehyde,acmc-1cof4,3-bromo-4-hydroxy-5-ethoxybenzaldehyde,3-bromo-5-ethoxy4-hydroxy-benzaldehyde,5-bromo-3-ethoxy-4-hydroxybenzaldehyde,3-bromo-5-ethoxy-4-hydroxy benzaldehyde,benzaldehyde,3-bromo-5-ethoxy-4-hydroxy,3-bromanyl-5-ethoxy-4-oxidanyl-benzaldehyde
612496
3-bromo-5-ethoxy-4-hydroxybenzaldehyde
CCOC1=C(C(=CC(=C1)C=O)Br)O
Sensitivity Air Sensitive
Quantity 1g
Melting Point 141°C to 143°C
Formula Weight 245.08
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-ethoxy-4-hydroxybenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures