Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-fluoro-2-methoxypyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
884494-81-9
C6H5BrFNO
206.014
MFCD04972397
HSWCOAGQYSBFAK-UHFFFAOYSA-N
3-bromo-5-fluoro-2-methoxypyrdine,2-methoxy-3-bromo-5-fluoropyridine,synquest 4h07-b-01,3-bromo-5-fluoro-2-methoxy-pyridine,pyridine,3-bromo-5-fluoro-2-methoxy,pyridine, 3-bromo-5-fluoro-2-methoxy,pubchem1175,acmc-209qt8,3-bromo-5-fluoropyridin-2-yl methyl ether,3-bromo-5-fluoro-2-methoxy pyridine
40427006
3-bromo-5-fluoro-2-methoxypyridine
COC1=C(C=C(C=N1)F)Br
884494-81-9
C6H5BrFNO
206.014
MFCD04972397
HSWCOAGQYSBFAK-UHFFFAOYSA-N
3-bromo-5-fluoro-2-methoxypyrdine,2-methoxy-3-bromo-5-fluoropyridine,synquest 4h07-b-01,3-bromo-5-fluoro-2-methoxy-pyridine,pyridine,3-bromo-5-fluoro-2-methoxy,pyridine, 3-bromo-5-fluoro-2-methoxy,pubchem1175,acmc-209qt8,3-bromo-5-fluoropyridin-2-yl methyl ether,3-bromo-5-fluoro-2-methoxy pyridine
40427006
3-bromo-5-fluoro-2-methoxypyridine
COC1=C(C=C(C=N1)F)Br
Quantity 5g
Sensitivity Light Sensitive
Beilstein 15497859
Formula Weight 206.01
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-fluoro-2-methoxypyridine

storageNote1 : 2° to 8°C