Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluorobenzonitrile, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
179898-34-1
C7H3BrFN
200.01
IADLVSLZPQYXIF-UHFFFAOYSA-N
3-fluoro-5-bromobenzonitrile,benzonitrile, 3-bromo-5-fluoro,5-bromo-3-fluorobenzenecarbonitrile,3-cyano-5-fluorobromobenzene,3-bromo 5-fluorobenzonitrile,3-bromo-5-fluoro-benzonitrile,3-bromo-5-fluorobenzenecarbonitrile,pubchem3220,acmc-209yem,3-fluoro-5-bromobenzonitile
2783330
3-bromo-5-fluorobenzonitrile
C1=C(C=C(C=C1F)Br)C#N
179898-34-1
C7H3BrFN
200.01
IADLVSLZPQYXIF-UHFFFAOYSA-N
3-fluoro-5-bromobenzonitrile,benzonitrile, 3-bromo-5-fluoro,5-bromo-3-fluorobenzenecarbonitrile,3-cyano-5-fluorobromobenzene,3-bromo 5-fluorobenzonitrile,3-bromo-5-fluoro-benzonitrile,3-bromo-5-fluorobenzenecarbonitrile,pubchem3220,acmc-209yem,3-fluoro-5-bromobenzonitile
2783330
3-bromo-5-fluorobenzonitrile
C1=C(C=C(C=C1F)Br)C#N
Quantity 5g
Melting Point 43.0°C to 46.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-fluorobenzonitrile

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning