Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluoropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
407-20-5
C5H3BrFN
175.99
HNNNBQRRIHKFLI-UHFFFAOYSA-N
5-bromo-3-fluoropyridine,3-bromo-5-fluoro-pyridine,3-brom-5-fluorpyridin,3-bromo-5-fluoro pyridine,pyridine, 3-bromo-5-fluoro,3-bromo-5-fluorpyridine,5-bromo-3-fluoride pyridine,3-bromine-5-fluoride pyridine,pubchem3964,3-bromo-5-fluoropyridin
820206
3-bromo-5-fluoropyridine
C1=C(C=NC=C1Br)F
407-20-5
C5H3BrFN
175.99
HNNNBQRRIHKFLI-UHFFFAOYSA-N
5-bromo-3-fluoropyridine,3-bromo-5-fluoro-pyridine,3-brom-5-fluorpyridin,3-bromo-5-fluoro pyridine,pyridine, 3-bromo-5-fluoro,3-bromo-5-fluorpyridine,5-bromo-3-fluoride pyridine,3-bromine-5-fluoride pyridine,pubchem3964,3-bromo-5-fluoropyridin
820206
3-bromo-5-fluoropyridine
C1=C(C=NC=C1Br)F
Melting Point 24.0°C to 28.0°C
Color Colorless to White
Flash Point 62°C
Boiling Point 78.0°C (11.0 Torr)
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 175.99
Physical Form Low Melting Solid or Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluoropyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger