Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-(hydroxymethyl)pyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
37669-64-0
C6H6BrNO
188.02
WDVDHJLKXYCOFS-UHFFFAOYSA-N
5-bromopyridin-3-yl methanol,5-bromo-3-pyridinemethanol,3-bromo-5-hydroxymethylpyridine,5-bromo-3-hydroxymethylpyridine,5-bromo-3-pyridinyl methanol,5-bromo-pyridin-3-yl-methanol,3-pyridinemethanol, 5-bromo,5-bromo-3-pyridyl methan-1-ol,3-bromo-5-hydroxymethyl pyridine,3-bromo-pyridin-5-yl methanol
2784733
(5-bromopyridin-3-yl)methanol
C1=C(C=NC=C1Br)CO
37669-64-0
C6H6BrNO
188.02
WDVDHJLKXYCOFS-UHFFFAOYSA-N
5-bromopyridin-3-yl methanol,5-bromo-3-pyridinemethanol,3-bromo-5-hydroxymethylpyridine,5-bromo-3-hydroxymethylpyridine,5-bromo-3-pyridinyl methanol,5-bromo-pyridin-3-yl-methanol,3-pyridinemethanol, 5-bromo,5-bromo-3-pyridyl methan-1-ol,3-bromo-5-hydroxymethyl pyridine,3-bromo-pyridin-5-yl methanol
2784733
(5-bromopyridin-3-yl)methanol
C1=C(C=NC=C1Br)CO
Packaging Glass Bottle
Boiling Point 154.0°C (4.0 Torr)
Quantity 1g
Color Yellow
Melting Point 27.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 188.02
Physical Form Liquid or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(hydroxymethyl)pyridine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning