Alfa Aesar™ 3-Bromo-5-hydroxypyridine, 95%

Manufacturer: Alfa Aesar™ H2725603

Quantity 1g

 View more versions of this product

Catalog No. AAH2725603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74115-13-2
    Molecular Formula C5H4BrNO
    Formula Weight 174
    MDL Number MFCD00661305
    Synonym 5-Bromo-3-pyridinol
    Chemical Name or Material 3-Bromo-5-hydroxypyridine

    Specifications

    Formula Weight 174
    Molecular Formula C5H4BrNO
    MDL Number MFCD00661305
    Synonym 5-Bromo-3-pyridinol
    Quantity 1g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 74115-13-2
    Chemical Name or Material 3-Bromo-5-hydroxypyridine
    Assay 95%

    Description

    Applications
    It is used in the total synthesis of (+-)-tabersonine.

    Solubility
    Slightly soluble in water. Soluble in chloroform and ethyl acetate.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit