Alfa Aesar™ 3-Bromo-5-methoxypyridine, 97%

Manufacturer: Alfa Aesar™ H2772803

Quantity 1g

 View more versions of this product

Catalog No. AAH2772803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50720-12-2
    Molecular Formula C6H6BrNO
    Formula Weight 188.02
    MDL Number MFCD00234169
    Chemical Name or Material 3-Bromo-5-methoxypyridine

    Specifications

    Formula Weight 188.02
    Molecular Formula C6H6BrNO
    MDL Number MFCD00234169
    CAS 50720-12-2
    Melting Point 29° to 33°C
    Quantity 1g
    Flash Point 102°C (216°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P264-P305+P351+P338
    Wear protective gloves.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Bromo-5-methoxypyridine
    Assay 97%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P264-P305+P351+P338
    Wear protective gloves.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit