Alfa Aesar™ 3-Bromo-5-methylbenzotrifluoride, 99%

Manufacturer: Alfa Aesar™ H3204003

Quantity 1g

 View more versions of this product

Catalog No. AAH3204003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86845-28-5
    Molecular Formula C8H6BrF3
    Formula Weight 239.04
    MDL Number MFCD01631585
    Synonym 3-Bromo-5-(trifluoromethyl)toluene; 5-Bromo-alpha,alpha,alpha-trifluoro-m-xylene
    Chemical Name or Material 3-Bromo-5-methylbenzotrifluoride

    Specifications

    Formula Weight 239.04
    Molecular Formula C8H6BrF3
    MDL Number MFCD01631585
    Synonym 3-Bromo-5-(trifluoromethyl)toluene; 5-Bromo-alpha,alpha,alpha-trifluoro-m-xylene
    Quantity 1g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    CAS 86845-28-5
    Chemical Name or Material 3-Bromo-5-methylbenzotrifluoride
    Assay 99%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit