Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-nitrobenzaldehyde, ≥97%

Click to view available options
Quantity:
250mg
1g
5g
355134-13-3
C7H4BrNO3
230.017
MFCD00100097
OUDCOMBHRXKPIJ-UHFFFAOYSA-N
benzaldehyde, 3-bromo-5-nitro,3-bromo-5-nitro-benzaldehyde,3-bromo-5-nitro benzaldehyde,ksc497m5f,3-nitro-5-bromo-benzaldehyde,3-bromanyl-5-nitro-benzaldehyde,5-bromo-3-nitrobenzaldehyde,3-bromo-5-nitrobenzaldehyde
16743966
3-bromo-5-nitrobenzaldehyde
C1=C(C=C(C=C1[N+](=O)[O-])Br)C=O
355134-13-3
C7H4BrNO3
230.017
MFCD00100097
OUDCOMBHRXKPIJ-UHFFFAOYSA-N
benzaldehyde, 3-bromo-5-nitro,3-bromo-5-nitro-benzaldehyde,3-bromo-5-nitro benzaldehyde,ksc497m5f,3-nitro-5-bromo-benzaldehyde,3-bromanyl-5-nitro-benzaldehyde,5-bromo-3-nitrobenzaldehyde,3-bromo-5-nitrobenzaldehyde
16743966
3-bromo-5-nitrobenzaldehyde
C1=C(C=C(C=C1[N+](=O)[O-])Br)C=O
Quantity 1g
Color Brown
Melting Point 101°C to 106°C
Beilstein 3253229
Formula Weight 230.02
Physical Form Powder
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-5-nitrobenzaldehyde

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning