Alfa Aesar™ 3-Bromo-5-nitrobenzotrifluoride, 97%

Manufacturer: Alfa Aesar™ H2617103

Quantity 1g

 View more versions of this product

Catalog No. AAH2617103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 630125-49-4
    Molecular Formula C7H3BrF3NO2
    Formula Weight 270.01
    MDL Number MFCD03788719
    Synonym 1-Bromo-3-nitro-5-(trifluoromethyl)benzene
    Chemical Name or Material 3-Bromo-5-nitrobenzotrifluoride

    Specifications

    Formula Weight 270.01
    MDL Number MFCD03788719
    Quantity 1g
    Molecular Formula C7H3BrF3NO2
    Synonym 1-Bromo-3-nitro-5-(trifluoromethyl)benzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    CAS 630125-49-4
    Chemical Name or Material 3-Bromo-5-nitrobenzotrifluoride
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit