Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-nitrobenzotrifluoride, 97%

Manufacturer:  Alfa Aesar™ H2617106

Quantity 5g

 View more versions of this product

Catalog No. AAH2617106


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

630125-49-4
270.01
1-Bromo-3-nitro-5-(trifluoromethyl)benzene
C7H3BrF3NO2
MFCD03788719
3-Bromo-5-nitrobenzotrifluoride

Specifications

630125-49-4
270.01
1-Bromo-3-nitro-5-(trifluoromethyl)benzene
3-Bromo-5-nitrobenzotrifluoride
C7H3BrF3NO2
MFCD03788719
5g
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit