Alfa Aesar™ 3-Bromo-5-(trifluoromethoxy)aniline, 97%

Manufacturer: Alfa Aesar™ H3251003

Quantity 1g

 View more versions of this product

Catalog No. AAH3251003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 914636-35-4
    Molecular Formula C7H5BrF3NO
    Formula Weight 256.02
    MDL Number MFCD08458076
    UN Number UN2810
    Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)aniline

    Specifications

    Formula Weight 256.02
    Quantity 1g
    Molecular Formula C7H5BrF3NO
    MDL Number MFCD08458076
    CAS 914636-35-4
    UN Number UN2810
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)aniline
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit