Learn More
Alfa Aesar™ 3'-Bromo-5'-(trifluoromethyl)acetophenone, 97%
Manufacturer:  Alfa Aesar™ H31854MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH31854MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
C9H6BrF3O
267.04
MFCD11847339
3'-Bromo-5'-(trifluoromethyl)acetophenone
Specifications
C9H6BrF3O
MFCD11847339
3'-Bromo-5'-(trifluoromethyl)acetophenone
267.04
250mg
97%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning