Alfa Aesar™ 3-Bromo-6-chlorochromone, 96%

Manufacturer: Alfa Aesar™ H32041MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32041MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73220-38-9
    Molecular Formula C9H4BrClO2
    Formula Weight 259.49
    MDL Number MFCD02093123
    Synonym 3-Bromo-6-chloro-4H-chromen-4-one
    Chemical Name or Material 3-Bromo-6-chlorochromone

    Specifications

    Formula Weight 259.49
    Molecular Formula C9H4BrClO2
    MDL Number MFCD02093123
    CAS 73220-38-9
    Melting Point 128° to 130°C
    Synonym 3-Bromo-6-chloro-4H-chromen-4-one
    Quantity 250mg
    Chemical Name or Material 3-Bromo-6-chlorochromone
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit