Learn More
eMolecules​ 3-BROMO-6-METHYLPYRIDAZINE | 65202-58-6 | 1G | Purity: 97% DFS Item
Manufacturer:  eMolecules​ 248476071
AstaTech | 3-BROMO-6-METHYLPYRIDAZINE | 1G | 65202-58-6 | MFCD09831907
Catalog No. 50-157-8578
Due to product restrictions, please to purchase or view availability for this product.