Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-7-azaindole, 96%

Click to view available options
Quantity:
1g
5g
74420-15-8
C7H5BrN2
197.035
MFCD02179276
VJDGIJDCXIEXPF-UHFFFAOYSA-N
3-bromo-7-azaindole,3-bromo-1h-pyrrolo 2,3-b pyridine,3-bromo-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 3-bromo,3-bromo-7-azaindol,3-bromopyrrolo 2,3-b pyridine,pubchem14707,acmc-209oup,ksc494e0h,3-bromo-7-azaindole
5523659
3-bromo-1H-pyrrolo[2,3-b]pyridine
C1=CC2=C(NC=C2Br)N=C1
74420-15-8
C7H5BrN2
197.035
MFCD02179276
VJDGIJDCXIEXPF-UHFFFAOYSA-N
3-bromo-7-azaindole,3-bromo-1h-pyrrolo 2,3-b pyridine,3-bromo-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 3-bromo,3-bromo-7-azaindol,3-bromopyrrolo 2,3-b pyridine,pubchem14707,acmc-209oup,ksc494e0h,3-bromo-7-azaindole
5523659
3-bromo-1H-pyrrolo[2,3-b]pyridine
C1=CC2=C(NC=C2Br)N=C1
Color White to Brown
Quantity 5g
Melting Point 185°C to 189°C
UN Number UN2811
Beilstein 119335
Formula Weight 197.04
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 3-Bromo-7-azaindole

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.