Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-7-nitroindazole, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

74209-34-0
C7H4BrN3O2
242.032
NFSTZPMYAZRZPC-UHFFFAOYSA-N
1649

Selective nNOS inhibitor

Potent inhibitor of rat cerebellar nitric oxide synthase; more potent than 7-nitroindazole (Cat. No. 0602).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C1=CC2=C(NN=C2C(=C1)[N+](=O)[O-])Br
IUPAC Name 3-bromo-7-nitro-2H-indazole
Formula Weight 242.03
Chemical Name or Material 3-Bromo-7-nitroindazole
Grade >98%

Recommended Storage : Desiccate at +4°C