Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-DL-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
30163-20-3
C9H10BrNO2
244.088
MFCD05227935
GDMOHOYNMWWBAU-UHFFFAOYSA-N
2-amino-3-3-bromophenyl propanoic acid,3-bromo-dl-phenylalanine,3-bromophenylalanine,dl-3-br-phe-oh,h-dl-phe 3-br-oh,dl-3-bromophenylalanine,pubchem18643,3-bromophenylalanine #,2-3-bromobenzyl glycine,dl-3-br-phe
554078
2-amino-3-(3-bromophenyl)propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)N
30163-20-3
C9H10BrNO2
244.088
MFCD05227935
GDMOHOYNMWWBAU-UHFFFAOYSA-N
2-amino-3-3-bromophenyl propanoic acid,3-bromo-dl-phenylalanine,3-bromophenylalanine,dl-3-br-phe-oh,h-dl-phe 3-br-oh,dl-3-bromophenylalanine,pubchem18643,3-bromophenylalanine #,2-3-bromobenzyl glycine,dl-3-br-phe
554078
2-amino-3-(3-bromophenyl)propanoic acid
C1=CC(=CC(=C1)Br)CC(C(=O)O)N
Beilstein 2939808
Formula Weight 244.09
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material 3-Bromo-DL-phenylalanine