Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromoaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
591-19-5
C6H6BrN
172.02
MFCD00007757
DHYHYLGCQVVLOQ-UHFFFAOYSA-N
m-bromoaniline,benzenamine, 3-bromo,3-bromo aniline,m-aminobromobenzene,aniline, m-bromo,3-bromobenzenamine,3-bromophenylamine,bromoaniline 3-,3-bromo-benzenamine,3-bromo-phenylamine
11562
3-bromoaniline
C1=CC(=CC(=C1)Br)N
591-19-5
C6H6BrN
172.02
MFCD00007757
DHYHYLGCQVVLOQ-UHFFFAOYSA-N
m-bromoaniline,benzenamine, 3-bromo,3-bromo aniline,m-aminobromobenzene,aniline, m-bromo,3-bromobenzenamine,3-bromophenylamine,bromoaniline 3-,3-bromo-benzenamine,3-bromo-phenylamine
11562
3-bromoaniline
C1=CC(=CC(=C1)Br)N
Flash Point >230°C
Melting Point 16.8°C
Specific Gravity 1.58
Boiling Point 251°C
Density 1.5800g/mL
Refractive Index 1.6240 to 1.626
Quantity 5mL
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H4NH2
Beilstein 12, 633
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents
Formula Weight 172.02
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Bromoaniline
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-704-4