Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromoanisole, 98+%

Click to view available options
Quantity:
25g
100g
500g
2398-37-0
C7H7BrO
187.036
MFCD00000081
PLDWAJLZAAHOGG-UHFFFAOYSA-N
3-bromoanisole,m-bromoanisole,3-methoxybromobenzene,3-bromo anisole,m-bromomethoxybenzene,benzene, 1-bromo-3-methoxy,anisole, m-bromo,3-methoxy-1-bromobenzene,m-methoxybromobenzene,meta-bromoanisole
16971
1-bromo-3-methoxybenzene
COC1=CC(=CC=C1)Br
2398-37-0
C7H7BrO
187.036
MFCD00000081
PLDWAJLZAAHOGG-UHFFFAOYSA-N
3-bromoanisole,m-bromoanisole,3-methoxybromobenzene,3-bromo anisole,m-bromomethoxybenzene,benzene, 1-bromo-3-methoxy,anisole, m-bromo,3-methoxy-1-bromobenzene,m-methoxybromobenzene,meta-bromoanisole
16971
1-bromo-3-methoxybenzene
COC1=CC(=CC=C1)Br
Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Refractive Index 1.564
Quantity 100g
Melting Point ~2°C
Density 1.489
Flash Point 93°C (199°F)
Boiling Point 210°C to 211°C
Beilstein 1524687
Solubility Information Miscible in methanol,benzene,hexane,toluene and dichloromethane. Practically insoluble in water.
Formula Weight 187.04
Percent Purity ≥98%
Chemical Name or Material 3-Bromoanisole
Show More Show Less

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 219-264-5

TSCA : Yes

Recommended Storage : Ambient temperatures