Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromobenzaldoxime, 97%

Click to view available options
Quantity:
1g
5g
25g
51873-95-1
C7H6BrNO
200.035
MFCD00143098
DBGBMIPNNIZQNN-WEVVVXLNSA-N
3-bromobenzaldoxime,3-bromobenzaldehyde oxime,e-3-bromobenzaldehyde oxime,pubchem3732,e-n-3-bromophenyl methylidene hydroxylamine,z-3-bromobenzaldehyde oxime,benzaldehyde, 3-bromo-, oxime,z-n-3-bromophenyl methylidene hydroxylamine,ne-n-3-bromophenyl methylidene hydroxylamine
5375574
(NE)-N-[(3-bromophenyl)methylidene]hydroxylamine
C1=CC(=CC(=C1)Br)C=NO
51873-95-1
C7H6BrNO
200.035
MFCD00143098
DBGBMIPNNIZQNN-WEVVVXLNSA-N
3-bromobenzaldoxime,3-bromobenzaldehyde oxime,e-3-bromobenzaldehyde oxime,pubchem3732,e-n-3-bromophenyl methylidene hydroxylamine,z-3-bromobenzaldehyde oxime,benzaldehyde, 3-bromo-, oxime,z-n-3-bromophenyl methylidene hydroxylamine,ne-n-3-bromophenyl methylidene hydroxylamine
5375574
(NE)-N-[(3-bromophenyl)methylidene]hydroxylamine
C1=CC(=CC(=C1)Br)C=NO
Melting Point 70°C to 74°C
Quantity 25g
Beilstein 2040559
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 3-Bromobenzaldoxime

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning