Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromobenzoyl chloride, 98%

Click to view available options
Quantity:
10g
50g
250g
1711-09-7
C7H4BrClO
219.462
MFCD00000669
PBOOZQFGWNZNQE-UHFFFAOYSA-N
m-bromobenzoyl chloride,benzoyl chloride, 3-bromo,3-bromobenzoylchloride,benzoyl chloride, m-bromo,3-bromobenzoic acid chloride,attercop-chm at106527,3-brom-benzoylchlorid,3-bromo-benzoylchlorid,3-bromobenzoic chloride,pubchem13574
74377
3-bromobenzoyl chloride
C1=CC(=CC(=C1)Br)C(=O)Cl
1711-09-7
C7H4BrClO
219.462
MFCD00000669
PBOOZQFGWNZNQE-UHFFFAOYSA-N
m-bromobenzoyl chloride,benzoyl chloride, 3-bromo,3-bromobenzoylchloride,benzoyl chloride, m-bromo,3-bromobenzoic acid chloride,attercop-chm at106527,3-brom-benzoylchlorid,3-bromo-benzoylchlorid,3-bromobenzoic chloride,pubchem13574
74377
3-bromobenzoyl chloride
C1=CC(=CC(=C1)Br)C(=O)Cl
Density 1.666
Refractive Index 1.595
Sensitivity Moisture sensitive
Quantity 10g
Boiling Point 247°C to 251°C
Flash Point 107°C (224°F)
UN Number UN3265
Beilstein 1635505
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 3-Bromobenzoyl chloride
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 216-978-9

TSCA : Yes

Recommended Storage : Ambient temperatures