Alfa Aesar™ 3-Bromobenzyl alcohol, 99%

Manufacturer: Alfa Aesar™ A10929-14

Quantity 25g

 View more versions of this product

Catalog No. AAA1092914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15852-73-0
    Molecular Formula C7H7BrO
    Formula Weight 187.04g/mol
    MDL Number MFCD00004629
    Beilstein 2242512
    Chemical Name or Material 3-Bromobenzyl alcohol

    Specifications

    Boiling Point 165°C (16mmHg)
    Quantity 25g
    Flash Point >110°C (230°F)
    CAS 15852-73-0
    Molecular Formula C7H7BrO
    Formula Weight 187.04g/mol
    MDL Number MFCD00004629
    Beilstein 2242512
    Chemical Name or Material 3-Bromobenzyl alcohol
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit