Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromobenzyl alcohol, 99%

Manufacturer:  Alfa Aesar™ A1092922

Quantity 100g

 View more versions of this product

Catalog No. AAA1092922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15852-73-0
187.04
2242512
C7H7BrO
MFCD00004629
3-Bromobenzyl alcohol

Specifications

15852-73-0
100g
C7H7BrO
MFCD00004629
3-Bromobenzyl alcohol
165°C (16mmHg)
>110°C (230°F)
187.04
2242512
99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit