Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromobenzylamine, 97%

Click to view available options
Quantity:
5g
25g
100g
10269-01-9
C7H8BrN
186.052
MFCD01026119
SUYJXERPRICYRX-UHFFFAOYSA-N
3-bromobenzylamine,3-bromophenyl methanamine,benzenemethanamine, 3-bromo,3-bromophenyl methylamine,m-bromobenzylamine,1-3-bromophenyl methanamine,3-bromo-benzylamine,3-bromobenzenemethanamine,3-bromobenzyl amine,pubchem21109
457587
(3-bromophenyl)methanamine
C1=CC(=CC(=C1)Br)CN
10269-01-9
C7H8BrN
186.052
MFCD01026119
SUYJXERPRICYRX-UHFFFAOYSA-N
3-bromobenzylamine,3-bromophenyl methanamine,benzenemethanamine, 3-bromo,3-bromophenyl methylamine,m-bromobenzylamine,1-3-bromophenyl methanamine,3-bromo-benzylamine,3-bromobenzenemethanamine,3-bromobenzyl amine,pubchem21109
457587
(3-bromophenyl)methanamine
C1=CC(=CC(=C1)Br)CN
Applications
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Quantity 100g
Odor Amine-like
UN Number UN2735
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromobenzylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.