Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobutyric acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
25g
2623-86-1
C4H7BrO2
167.002
HAIUIAZIUDPZIE-UHFFFAOYSA-N
3-bromobutyric acid,butanoic acid, 3-bromo,acmc-1cjov,#,3-bromo butyric acid,3-bromobutyric acid gc
102860
3-bromobutanoic acid
CC(CC(=O)O)Br
2623-86-1
C4H7BrO2
167.002
HAIUIAZIUDPZIE-UHFFFAOYSA-N
3-bromobutyric acid,butanoic acid, 3-bromo,acmc-1cjov,#,3-bromo butyric acid,3-bromobutyric acid gc
102860
3-bromobutanoic acid
CC(CC(=O)O)Br
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 167
Chemical Name or Material 3-Bromobutyric acid