Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+)-3-Bromocamphor, 98%

Click to view available options
Quantity:
100g
500g
10293-06-8
C10H15BrO
231.133
MFCD00003744
NJQADTYRAYFBJN-MZVBAPTMSA-N
endo-3-bromo-d-camphor,+-camphor bromide,3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
44630109
(1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC1(C(=O)C2Br)C)C
10293-06-8
C10H15BrO
231.133
MFCD00003744
NJQADTYRAYFBJN-MZVBAPTMSA-N
endo-3-bromo-d-camphor,+-camphor bromide,3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
44630109
(1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC1(C(=O)C2Br)C)C
Sensitivity Light Sensitive
Melting Point 75°C to 77°C
Quantity 100g
Color White
Beilstein 3197237
Formula Weight 231.13
Physical Form Crystalline
Percent Purity 98%
Chemical Name or Material (+)-3-Bromocamphor

RTECSNumber : ED6750000