Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromocarbazole, 98%

Click to view available options
Quantity:
1g
5g
25g
1592-95-6
C12H8BrN
246.107
MFCD00222621
LTBWKAYPXIIVPC-UHFFFAOYSA-N
3-bromocarbazole,9h-carbazole, 3-bromo,3-brominecarbazole,3bromo-9h-carbazole,pubchem9950,maybridge1_004311,acmc-1bpt1,3-bromocarbazole gc
252446
3-bromo-9H-carbazole
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br
1592-95-6
C12H8BrN
246.107
MFCD00222621
LTBWKAYPXIIVPC-UHFFFAOYSA-N
3-bromocarbazole,9h-carbazole, 3-bromo,3-brominecarbazole,3bromo-9h-carbazole,pubchem9950,maybridge1_004311,acmc-1bpt1,3-bromocarbazole gc
252446
3-bromo-9H-carbazole
C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br
Applications
Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Melting Point 195°C to 199°C
Color White
Quantity 25g
Boiling Point 160°C (6mmHg)
Beilstein 142195
Formula Weight 246.11
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Bromocarbazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning