Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromochlorobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
108-37-2
C6H4BrCl
191.45
MFCD00000568
JRGGUPZKKTVKOV-UHFFFAOYSA-N
3-bromochlorobenzene,m-bromochlorobenzene,3-chlorobromobenzene,benzene, 1-bromo-3-chloro,m-chlorobromobenzene,1-chloro-3-bromobenzene,3-chlorophenyl bromide,m-chlorophenyl bromide,m-bromophenyl chloride,3-bromo-1-chlorobenzene
7928
1-bromo-3-chlorobenzene
C1=CC(=CC(=C1)Br)Cl
108-37-2
C6H4BrCl
191.45
MFCD00000568
JRGGUPZKKTVKOV-UHFFFAOYSA-N
3-bromochlorobenzene,m-bromochlorobenzene,3-chlorobromobenzene,benzene, 1-bromo-3-chloro,m-chlorobromobenzene,1-chloro-3-bromobenzene,3-chlorophenyl bromide,m-chlorophenyl bromide,m-bromophenyl chloride,3-bromo-1-chlorobenzene
7928
1-bromo-3-chlorobenzene
C1=CC(=CC(=C1)Br)Cl
Infrared Spectrum Authentic
Quantity 25mL
Flash Point 80°C
Specific Gravity 1.63
Melting Point -21.5°C
Refractive Index 1.5760 to 1.5780
Density 1.6300g/mL
Packaging Glass bottle
Boiling Point 195.0°C to 197.0°C
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula BrC6H4Cl
Beilstein 05, 209
Solubility Information Solubility in water: insoluble
Formula Weight 191.45
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Bromochlorobenzene, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 203-575-8

RTECSNumber : CY9005000

TSCA : TSCA