Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocinnamic Acid, Predominantly trans 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Plastic bottle
32862-97-8
C9H7BrO2
227.057
MFCD00004382
YEMUSDCFQUBPAL-SNAWJCMRSA-N
3-bromocinnamic acid,3-3-bromophenyl acrylic acid,m-bromocinnamic acid,trans-3-bromocinnamic acid,cinnamic acid, m-bromo,bw a800u,e-3-3-bromophenyl acrylic acid,3-3-bromophenyl-2-propenoic acid,2e-3-3-bromophenyl acrylic acid,bromocinnamic acid,3
776461
(E)-3-(3-bromophenyl)prop-2-enoic acid
C1=CC(=CC(=C1)Br)C=CC(=O)O
32862-97-8
C9H7BrO2
227.057
MFCD00004382
YEMUSDCFQUBPAL-SNAWJCMRSA-N
3-bromocinnamic acid,3-3-bromophenyl acrylic acid,m-bromocinnamic acid,trans-3-bromocinnamic acid,cinnamic acid, m-bromo,bw a800u,e-3-3-bromophenyl acrylic acid,3-3-bromophenyl-2-propenoic acid,2e-3-3-bromophenyl acrylic acid,bromocinnamic acid,3
776461
(E)-3-(3-bromophenyl)prop-2-enoic acid
C1=CC(=CC(=C1)Br)C=CC(=O)O
Infrared Spectrum Authentic
Quantity 25g
Packaging Plastic bottle
Color White to Yellow
Melting Point 178°C to 182°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4CH=CHCO2H
Beilstein 09, 597
Formula Weight 227.06
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Bromocinnamic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 251-267-7

RTECSNumber : GD8420000

TSCA : TSCA