Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocumene 98.0+%, TCI America™
SDP

Catalog No. p-7119821
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5433-01-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD01318112
InChI Key GBSGGFCCQZUXNB-UHFFFAOYSA-N
Synonym 1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene
PubChem CID 138499
IUPAC Name 1-bromo-3-propan-2-ylbenzene
SMILES CC(C)C1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 199.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromocumene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.