Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocyclohexene, 95%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
1521-51-3
C6H9Br
161.04
MFCD00013775
AJKDUJRRWLQXHM-UHFFFAOYSA-N
3-bromocyclohex-1-ene,cyclohexene, 3-bromo,1-bromo-2-cyclohexene,3-bromo-1-cyclohexene,1,2,3,4-tetrahydrobromobenzene,3-bromo-cyclohexene,2-cyclohexenyl bromide,2-cyclohexen-1-yl bromide,3-bromo cyclohexene,3-bromanylcyclohexene
137057
3-bromocyclohexene
C1CC=CC(C1)Br
1521-51-3
C6H9Br
161.04
MFCD00013775
AJKDUJRRWLQXHM-UHFFFAOYSA-N
3-bromocyclohex-1-ene,cyclohexene, 3-bromo,1-bromo-2-cyclohexene,3-bromo-1-cyclohexene,1,2,3,4-tetrahydrobromobenzene,3-bromo-cyclohexene,2-cyclohexenyl bromide,2-cyclohexen-1-yl bromide,3-bromo cyclohexene,3-bromanylcyclohexene
137057
3-bromocyclohexene
C1CC=CC(C1)Br
Specific Gravity 1.38
Refractive Index 1.5280 to 1.5320
Boiling Point 64.0°C to 65.0°C (15.0mmHg)
Color White
Packaging Glass bottle
Quantity 250mL
Infrared Spectrum Authentic
Flash Point 54°C
Density 1.3800g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 161.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Bromocyclohexene
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

Recommended Storage : May darken on ageing