Learn More
eMolecules​ 3-Bromoindazole | 40598-94-5 | 1G | Purity: 95% DFS Item
Manufacturer:  eMolecules​ 389722698
Matrix Scientific | 3-Bromoindazole | 1G | 40598-94-5 | MFCD00159926
Catalog No. 50-157-7334
Due to product restrictions, please to purchase or view availability for this product.