Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-bromoindole-2-carboxylic Acid, +96%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
28737-33-9
C9H6BrNO2
240.056
MFCD02710285
KOTZNHLNBDTYMS-UHFFFAOYSA-N
3-bromoindole-2-carboxylic acid,1h-indole-2-carboxylicacid, 3-bromo,1h-indole-2-carboxylic acid, 3-bromo,enamine_001100,acmc-1cb22,ksc206a8j
2325688
3-bromo-1H-indole-2-carboxylic acid
C1=CC=C2C(=C1)C(=C(N2)C(=O)O)Br
28737-33-9
C9H6BrNO2
240.056
MFCD02710285
KOTZNHLNBDTYMS-UHFFFAOYSA-N
3-bromoindole-2-carboxylic acid,1h-indole-2-carboxylicacid, 3-bromo,1h-indole-2-carboxylic acid, 3-bromo,enamine_001100,acmc-1cb22,ksc206a8j
2325688
3-bromo-1H-indole-2-carboxylic acid
C1=CC=C2C(=C1)C(=C(N2)C(=O)O)Br
Quantity 250mg
Packaging Glass bottle
Color Beige-Pink
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96+%
Formula Weight 240.06
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Bromoindole-2-carboxylic acid
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.