Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals

Catalog Number ac397140010
Click to view available options
Quantity:
1 g
5 g
4916-55-6
C6H6BrN·HBr
252.93
MFCD01321314
FNHPUOJKUXFUKN-UHFFFAOYSA-N
3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide
12707037
3-(bromomethyl)pyridine;hydrobromide
C1=CC(=CN=C1)CBr.Br
4916-55-6
C6H6BrN·HBr
252.93
MFCD01321314
FNHPUOJKUXFUKN-UHFFFAOYSA-N
3-bromomethyl pyridine hydrobromide, 3-picolyl bromide hydrobromide, 3-bromomethylpyridine hydrobromide, 3-bromomethyl-pyridine hydrobromide, pyridine, 3-bromomethyl-, hydrobromide, pubchem17664, acmc-209kf1, 3-brommethyl pyridine-hbr, 3-bromomethyl pyridine, bromide, 3-bromomethyl pyridinehydrobromide
12707037
3-(bromomethyl)pyridine;hydrobromide
C1=CC(=CN=C1)CBr.Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150.0°C to 155.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 252.93
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)pyridine hydrobromide
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit