Learn More
3-Bromopentane, 90%, ACROS Organics™
Specifications
Color | Yellow |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Quantity | 25g |
Refractive Index | 1.4420 to 1.4460 (20°C, 589nm) |
Density | 1.21g/mL |
CAS Min % | 88.0 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Linear Formula | C2H5CH(Br)C2H5 |
Show More |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger