Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopentane, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1809-10-5
C5H11Br
151.047
MFCD00000158
VTOQFOCYBTVOJZ-UHFFFAOYSA-N
pentane, 3-bromo,3-pentyl bromide,3-pentylbromide,3-bromo-pentane,3-bromo pentane,3-bromopentane,acmc-1c3bs
15738
3-bromopentane
CCC(CC)Br
1809-10-5
C5H11Br
151.047
MFCD00000158
VTOQFOCYBTVOJZ-UHFFFAOYSA-N
pentane, 3-bromo,3-pentyl bromide,3-pentylbromide,3-bromo-pentane,3-bromo pentane,3-bromopentane,acmc-1c3bs
15738
3-bromopentane
CCC(CC)Br
Quantity 100g
Color Yellow
Packaging Glass bottle
Refractive Index 1.4420 to 1.4460 (20°C, 589nm)
Infrared Spectrum Authentic
Density 1.21g/mL
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula C2H5CH(Br)C2H5
Formula Weight 151.05
Physical Form Liquid
Percent Purity ≥88% (typically), 10% (2-Bromopentane) typically
Chemical Name or Material 3-Bromopentane, Containing about 10% 2-Bromopentane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger