Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenethyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
28229-69-8
C8H9BrO
201.06
MFCD00191857
PTTFLKHCSZSFOL-UHFFFAOYSA-N
3-bromophenethyl alcohol,2-3-bromophenyl ethanol,2-3-bromophenyl ethan-1-ol,benzeneethanol, 3-bromo,3-bromophenethylalcohol,3-bromobenzeneethanol,3-2-hydroxyethyl bromobenzene,2-3-bromophenyl ethyl alcohol,pubchem14821,3-bromo-phenethyl-alcohol
2734090
2-(3-bromophenyl)ethanol
C1=CC(=CC(=C1)Br)CCO
28229-69-8
C8H9BrO
201.06
MFCD00191857
PTTFLKHCSZSFOL-UHFFFAOYSA-N
3-bromophenethyl alcohol,2-3-bromophenyl ethanol,2-3-bromophenyl ethan-1-ol,benzeneethanol, 3-bromo,3-bromophenethylalcohol,3-bromobenzeneethanol,3-2-hydroxyethyl bromobenzene,2-3-bromophenyl ethyl alcohol,pubchem14821,3-bromo-phenethyl-alcohol
2734090
2-(3-bromophenyl)ethanol
C1=CC(=CC(=C1)Br)CCO
Specific Gravity 1.478
Flash Point >110°C
Refractive Index 1.5730 to 1.575
Boiling Point 107°C to 110°C (1.0mmHg)
Density 1.4780g/mL
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 201.06
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromophenethyl alcohol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning