Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenol, 98%, Acros Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
591-20-8
C6H5BrO
173.01
MFCD00002253
MNOJRWOWILAHAV-UHFFFAOYSA-N
m-bromophenol,phenol, 3-bromo,3-bromo phenol,3-bromo-phenol,phenol, m-bromo,unii-vmu0x6956y,meta bromophenol,meta-bromophenol,3 bromophenol,3-bromanylphenol
11563
3-bromophenol
C1=CC(=CC(=C1)Br)O
591-20-8
C6H5BrO
173.01
MFCD00002253
MNOJRWOWILAHAV-UHFFFAOYSA-N
m-bromophenol,phenol, 3-bromo,3-bromo phenol,3-bromo-phenol,phenol, m-bromo,unii-vmu0x6956y,meta bromophenol,meta-bromophenol,3 bromophenol,3-bromanylphenol
11563
3-bromophenol
C1=CC(=CC(=C1)Br)O
Flash Point >110°C
Infrared Spectrum Authentic
Quantity 100g
Melting Point 30.0°C to 32.0°C
Color Brown or Yellow
Refractive Index 1.5950 to 1.5990
Boiling Point 236.0°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4OH
Beilstein 06,198
Merck Index 15,1439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies,alcohol,ether,acetone
Formula Weight 173.01
Physical Form Liquid
Percent Purity 97.5 % min. (GC)
Chemical Name or Material 3-Bromophenol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.