Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
89598-96-9
C6H6BBrO2
200.83
MFCD00239386
AFSSVCNPDKKSRR-UHFFFAOYSA-N
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
2734318
(3-bromophenyl)boronic acid
B(C1=CC(=CC=C1)Br)(O)O
89598-96-9
C6H6BBrO2
200.83
MFCD00239386
AFSSVCNPDKKSRR-UHFFFAOYSA-N
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
2734318
(3-bromophenyl)boronic acid
B(C1=CC(=CC=C1)Br)(O)O
Melting Point 175°C to 177°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4B(OH)2
Solubility Information Solubility in water: 25g/L
Formula Weight 200.83
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Bromophenylboronic acid
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning