Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopropene, Stabilized with ca. 300 ppm Propylene Oxide, Approx. 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
200 L
Packaging:
Amber Glass Bottle
Drum
106-95-6
106-95-6
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Percent Purity Approx. 99%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.