Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-bromopropionaldehyde dimethylacetal, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
36255-44-4
C5H11BrO2
183.045
MFCD00013245
ODZZAIFAQLODKN-UHFFFAOYSA-N
3-bromopropionaldehyde dimethyl acetal,1-bromo-3,3-dimethoxypropane,propane, 3-bromo-1,1-dimethoxy,3-bromo-1,1-dimethoxy-propane,acmc-209ilq,3,3-dimethoxypropyl bromide,3-bromopropanal dimethylacetal,ksc495q3l,3-bromo-1,1dimethoxy-propane,1,1-dimethoxy-3-bromopropane
118932
3-bromo-1,1-dimethoxypropane
COC(CCBr)OC
36255-44-4
C5H11BrO2
183.045
MFCD00013245
ODZZAIFAQLODKN-UHFFFAOYSA-N
3-bromopropionaldehyde dimethyl acetal,1-bromo-3,3-dimethoxypropane,propane, 3-bromo-1,1-dimethoxy,3-bromo-1,1-dimethoxy-propane,acmc-209ilq,3,3-dimethoxypropyl bromide,3-bromopropanal dimethylacetal,ksc495q3l,3-bromo-1,1dimethoxy-propane,1,1-dimethoxy-3-bromopropane
118932
3-bromo-1,1-dimethoxypropane
COC(CCBr)OC
Refractive Index 1.4460 to 1.4500 (20°C, 589nm)
Packaging Glass bottle
Boiling Point 58.0°C to 60.0°C (14.0mmHg)
Color Colorless to Yellow
Density 1.3410g/mL
Infrared Spectrum Authentic
Flash Point 46°C
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula Br(CH2)2CH(OCH3)2
Formula Weight 183.04
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-Bromopropionaldehyde dimethylacetal
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning