Alfa Aesar™ 3-Bromopropionyl chloride, tech. 90%

Manufacturer: Alfa Aesar™ A1084622

Quantity 100g

 View more versions of this product

Catalog No. AAA1084622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 15486-96-1
    Molecular Formula C3H4BrClO
    Formula Weight 171.43
    MDL Number MFCD00000746
    UN Number UN3265
    Beilstein 605453
    Chemical Name or Material 3-Bromopropionyl chloride
    Grade Technical Grade

    Specifications

    Formula Weight 171.43
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00000746
    CAS 15486-96-1
    UN Number UN3265
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    TSCA TSCA
    Beilstein 605453
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula C3H4BrClO
    Boiling Point 54° to 56°C (14mmHg)
    Quantity 100g
    Flash Point 79°C (174°F)
    Chemical Name or Material 3-Bromopropionyl chloride
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    3-Bromopropionyl chloride acts as an acrylate synthon by reaction with 2-oxazolidinone, followed by base-promoted elimination to form a Diels-Alder dienophile. Brominated poly (ethylene glycol) methacrylate (v-PEG-Br) which was synthesized by the reaction of poly (ethyleneglycol) methacrylate, v-PEG-OH with Mn = 526 Dalton and 3-bromopropionyl chloride. Used in the reduction of organic compounds by mixed hydrides. Used as reagent in Chroman and tetrahydroquinoline ureas as potent TRPV1 antagonists. Reaction of veratrole with 3- bromopropionyl chloride led to the formation of 15 in 82% yield, which on treatment with the Tebbe reagent13 led to the formation of styryl bromide 16 in 93% yield. Used in the preparation of ome new mono- and di-nitrogen-containing monobenzo-crowns. 3-Bromopropionyl chloride was used in the preparation of 1-chloro-4-bromo-2-butanone.

    Solubility
    Reacts with water.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit