Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Bromopropoxy)-tert-butyldimethylsilane, 97%, stabilized over sodium carbonate, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
89031-84-5,497-19-8
C9H21BrOSi
253.26
QGMROEZDWJTIDW-UHFFFAOYSA-N
3-bromopropoxy-tert-butyldimethylsilane,3-bromopropoxy tert-butyl dimethylsilane,3-bromopropoxy-t-butyldimethylsilane,3-bromopropoxy-tert-butyl-dimethyl-silane,silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl,acmc-209unq,t-butyl 3-bromopropoxy dimethylsilane,bromopropoxy-tert-butyldimethyl silane
607486
3-bromopropoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCCBr
89031-84-5,497-19-8
C9H21BrOSi
253.26
QGMROEZDWJTIDW-UHFFFAOYSA-N
3-bromopropoxy-tert-butyldimethylsilane,3-bromopropoxy tert-butyl dimethylsilane,3-bromopropoxy-t-butyldimethylsilane,3-bromopropoxy-tert-butyl-dimethyl-silane,silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl,acmc-209unq,t-butyl 3-bromopropoxy dimethylsilane,bromopropoxy-tert-butyldimethyl silane
607486
3-bromopropoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCCBr
Specific Gravity 1.093
Quantity 5mL
Density 1.0930g/mL
Flash Point 85°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 182.0°C
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 253.26
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (3-Bromopropoxy)-tert-butyldimethylsilane, Stabilized
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning