Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.
Solubility
Soluble in water (50 mg/ml).
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.
Solubility
Soluble in water (50 mg/ml).
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications
Melting Point | 172°C to 174°C |
Quantity | 100 g |
Beilstein | 3906418 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water (50mg/ml). |
Formula Weight | 218.93 |
Percent Purity | 98% |
Chemical Name or Material | 3-Bromopropylamine hydrobromide |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 225-675-0 |
RTECSNumber | UH9925000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction