Alfa Aesar™ 3-Bromopropylboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H30646-14

Quantity 25g

 View more versions of this product

Catalog No. AAH3064614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 124215-44-7
    Molecular Formula C9H18BBrO2
    Formula Weight 248.96
    MDL Number MFCD10567053
    Synonym 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 3-Bromopropylboronic acid pinacol ester

    Specifications

    Formula Weight 248.96
    MDL Number MFCD10567053
    Boiling Point 61° to 64°C (0.35mmHg)
    Molecular Formula C9H18BBrO2
    CAS 124215-44-7
    Synonym 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Bromopropylboronic acid pinacol ester
    Assay 98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit