Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol

Click to view available options
Quantity:
1g
5g
25g
2039-86-3
C8H7Br
183.048
MFCD00000088
KQJQPCJDKBKSLV-UHFFFAOYSA-N
3-bromostyrene,1-bromo-3-vinylbenzene,m-bromostyrene,benzene, 1-bromo-3-ethenyl,1-bromo-3-vinyl-benzene,unii-6xe3sf241y,3-bromo-styrene,pubchem23859,acmc-1cmdc,1-bromo-3-vinylbenzene #
74870
1-bromo-3-ethenylbenzene
C=CC1=CC(=CC=C1)Br
2039-86-3
C8H7Br
183.048
MFCD00000088
KQJQPCJDKBKSLV-UHFFFAOYSA-N
3-bromostyrene,1-bromo-3-vinylbenzene,m-bromostyrene,benzene, 1-bromo-3-ethenyl,1-bromo-3-vinyl-benzene,unii-6xe3sf241y,3-bromo-styrene,pubchem23859,acmc-1cmdc,1-bromo-3-vinylbenzene #
74870
1-bromo-3-ethenylbenzene
C=CC1=CC(=CC=C1)Br
Quantity 1g
Flash Point 67°C (152°F)
Boiling Point 73°C to 74°C (3mmHg)
Beilstein 2038490
Formula Weight 183.05
Percent Purity 97%
Chemical Name or Material 3-Bromostyrene, stabilized with 0.1% 4-tert-butylcatechol

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C